3-Methylazetidine hydrochloride 95%

Lieferant: Apollo Scientific

OR309000-1G
APOSOR309000-1GEA 0 EUR
APOSOR309000-1G
3-Methylazetidine hydrochloride 95%
3-Methylazetidine hydrochloride
Formel: C4H10ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD09743448
CAS-Nummer: 935669-28-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR