(S)-1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid 97%

Lieferant: Apollo Scientific

OR309015-1G OR309015-10G OR309015-5G
APOSOR309015-1GEA 0 EUR
APOSOR309015-1G APOSOR309015-10G APOSOR309015-5G
(S)-1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid 97%
(S)-1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid
Formel: C₉H₁₅NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD01321011
CAS-Nummer: 51077-14-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR