Azetidine-2-carboxylic acid 98%

Lieferant: Apollo Scientific

OR309022-5G
APOSOR309022-5GEA 0 EUR
APOSOR309022-5G
Azetidine-2-carboxylic acid 98%
Azetidine-2-carboxylic acid
Formel: C₄H₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00066660
CAS-Nummer: 2517-04-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR