Azetidin-3-ylmethanamine dihydrochloride 97%

Lieferant: Apollo Scientific

OR309035-100MG
APOSOR309035-100MGEA 0 EUR
APOSOR309035-100MG
Azetidin-3-ylmethanamine dihydrochloride 97%
Azetidin-3-ylmethanamine dihydrochloride
Formel: C₄H₁₂Cl₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10565803
CAS-Nummer: 221095-80-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR