Benzyl-3-oxoazetidine-1-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

OR309040-100G OR309040-25G OR309040-5G
APOSOR309040-100GEA 261 EUR
APOSOR309040-100G APOSOR309040-25G APOSOR309040-5G
Benzyl-3-oxoazetidine-1-carboxylate 97%
Benzyl-3-oxoazetidine-1-carboxylate
Formel: C₁₁H₁₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09954923
CAS-Nummer: 105258-93-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR