1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid 95%

Lieferant: Apollo Scientific

OR309044-1G OR309044-5G
APOSOR309044-1GEA 63.8 EUR
APOSOR309044-1G APOSOR309044-5G
1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid 95%
1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid
Formel: C₉H₁₅NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD01861756
CAS-Nummer: 159749-28-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR