Azetidine-3-carbonitrile hydrochloride 95%

Lieferant: Apollo Scientific
Gefahr

OR309045-1G OR309045-5G
APOSOR309045-1GEA 0 EUR
APOSOR309045-1G APOSOR309045-5G
Azetidine-3-carbonitrile hydrochloride 95%
Azetidine-3-carbonitrile hydrochloride
Formel: C₄H₇ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09749828
CAS-Nummer: 345954-83-8
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR