Oxetane-3-carbaldehyde

Lieferant: Apollo Scientific
Achtung

OR309108-1G
APOSOR309108-1GEA 596 EUR
APOSOR309108-1G
Oxetane-3-carbaldehyde
Oxetane-3-carbaldehyde
Formel: C₄H₆O₂
Molecular Weight: 86,089 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD21604173
CAS-Nummer: 1305207-52-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR