5-(2-Aminoethyl)-1-methyl-1H-indazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(1-Methyl-1H-indazol-5-yl)ethylamine
OR30912-50MG
APOSOR30912-50MGEA
116
EUR
APOSOR30912-50MG
5-(2-Aminoethyl)-1-methyl-1H-indazole
5-(2-Aminoethyl)-1-methyl-1H-indazole
Formel:
C₁₀H₁₃N₃ Molecular Weight: 175,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12028591 CAS-Nummer: 1130309-67-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...