2-Oxa-6-azaspiro[3.5]nonane-6-carboxylic acid tert-butyl ester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 2-oxa-6-azaspiro[3.5]nonane-6-carboxylate

OR309123-1G
APOSOR309123-1GEA 184 EUR
APOSOR309123-1G
2-Oxa-6-azaspiro[3.5]nonane-6-carboxylic acid tert-butyl ester 97%
2-Oxa-6-azaspiro[3.5]nonane-6-carboxylic acid tert-butyl ester
Formel: C₁₂H₂₁NO₃
Molecular Weight: 227,3 g/mol
Schmelzpunkt: 74…77 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD12545892
CAS-Nummer: 1245816-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR