1-Oxa-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester 95+%
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 1-oxa-7-azaspiro[3.5]nonane-7-carboxylate
OR309130-1G
APOSOR309130-1GEA
139
EUR
APOSOR309130-1G
1-Oxa-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester 95+%
1-Oxa-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester
Formel:
C₁₂H₂₁NO₃ Molecular Weight: 227,3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08706105 CAS-Nummer: 864684-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...