4-Keto-L-proline hydrobromide 90%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Oxo-L-proline hydrobromide

OR309140-1G
APOSOR309140-1GEA 191 EUR
APOSOR309140-1G
4-Keto-L-proline hydrobromide 90%
4-Keto-L-proline hydrobromide
Formel: C₅H₈BrNO₃
Molecular Weight: 210.03 g/mol
Schmelzpunkt: 134…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12024572
CAS-Nummer: 75776-67-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR