tert-Butyl-3-bromopyrrolidine-1-carboxylate 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(±)-1-Boc-3-bromopyrrolidine
OR309142-5G
APOSOR309142-5GEA
87
EUR
APOSOR309142-5G
tert-Butyl-3-bromopyrrolidine-1-carboxylate 98%
tert-Butyl-3-bromopyrrolidine-1-carboxylate
Formel:
C₉H₁₆BrNO₂ Molecular Weight: 250.14 g/mol Schmelzpunkt: 47…52 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08752478 CAS-Nummer: 939793-16-5 UN: 3077 ADR: 9+EHS III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...