(R)-(-)-1-Boc-3-bromopyrrolidine 97%

Lieferant: Apollo Scientific

Synonyme: (R)-1-Boc-3-bromopyrrolidine

OR309143-250MG
APOSOR309143-250MGEA 71.2 EUR
APOSOR309143-250MG
(R)-(-)-1-Boc-3-bromopyrrolidine 97%
(R)-(-)-1-Boc-3-bromopyrrolidine
Formel: C₉H₁₆BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD17214727
CAS-Nummer: 569660-97-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR