(S)-(+)-1-Boc-3-bromopyrrolidine 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(S)-tert-Butyl 3-bromopyrrolidine-1-carboxylate
OR309144-1G
APOSOR309144-1GEA
387
EUR
APOSOR309144-1G
(S)-(+)-1-Boc-3-bromopyrrolidine 95%
(S)-(+)-1-Boc-3-bromopyrrolidine
Formel:
C₉H₁₆BrNO₂ Molecular Weight: 250,136 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD17214728 CAS-Nummer: 569660-89-5 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...