Ethyl pyrrolidine-3-carboxylate hydrochloride 95+%

Lieferant: Apollo Scientific

OR309148-5G OR309148-25G
APOSOR309148-5GEA 226 EUR
APOSOR309148-5G APOSOR309148-25G
Ethyl pyrrolidine-3-carboxylate hydrochloride 95+%
Ethyl pyrrolidine-3-carboxylate hydrochloride
Formel: C₇H₁₄ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD14585012
CAS-Nummer: 80028-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR