(S)-1-Boc-4-oxo-piperidine-2-carboxylic acid methyl ester 95+%

Lieferant: Apollo Scientific

OR309155-250MG
APOSOR309155-250MGEA 0 EUR
APOSOR309155-250MG
(S)-1-Boc-4-oxo-piperidine-2-carboxylic acid methyl ester 95+%
(S)-1-Boc-4-oxo-piperidine-2-carboxylic acid methyl ester
Formel: C₁₃H₂₁NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD15071881
CAS-Nummer: 180854-44-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR