(R)-Pyrrolidin-3-ylmethanol
Lieferant: Apollo Scientific
|
Achtung
|
OR309158-1G
APOSOR309158-1GEA
115
EUR
APOSOR309158-1G
(R)-Pyrrolidin-3-ylmethanol
(R)-Pyrrolidin-3-ylmethanol
Formel:
C₅H₁₁NO Molecular Weight: 101,15 g/mol Schmelzpunkt: 173…177 °C Dichte: 0,979 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD09607969 CAS-Nummer: 110013-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...