(R)-tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate 97%

Lieferant: Apollo Scientific

Synonyme: [R]-1-BOC-3-Pyrrolidinemethanol, (R)-tert-Butyl 3-(hydroxymethyl)pyrrolidine-1-carboxylate

OR309160-5G
APOSOR309160-5GEA 177 EUR
APOSOR309160-5G
(R)-tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate 97%
(R)-tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate
Formel: C₁₀H₁₉NO₃
Molecular Weight: 201,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094725
CAS-Nummer: 138108-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR