N-(tert-Butoxycarbonyl)-L-pyroglutamic acid 95+%

Lieferant: Apollo Scientific

OR309165-25G
APOSOR309165-25GEA 89.9 EUR
APOSOR309165-25G
N-(tert-Butoxycarbonyl)-L-pyroglutamic acid 95+%
N-(tert-Butoxycarbonyl)-L-pyroglutamic acid
Formel: C₁₀H₁₅NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00672316
CAS-Nummer: 53100-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR