N-Boc-trans-4-(fmoc-amino)-L-proline

Lieferant: Apollo Scientific
Achtung

OR309173-1G
APOSOR309173-1GEA 0 EUR
APOSOR309173-1G
N-Boc-trans-4-(fmoc-amino)-L-proline
N-Boc-trans-4-(fmoc-amino)-L-proline
Formel: C₂₅H₂₈N₂O₆
Schmelzpunkt: 199 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00673783
CAS-Nummer: 176486-63-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR