Ethyl-2-oxopyrrolidine-4-carboxylate 95+%

Lieferant: Apollo Scientific

Synonyme: Ethyl 5-oxopyrrolidine-3-carboxylate

OR309174-1G
APOSOR309174-1GEA 78.3 EUR
APOSOR309174-1G
Ethyl-2-oxopyrrolidine-4-carboxylate 95+%
Ethyl-2-oxopyrrolidine-4-carboxylate
Formel: C₇H₁₁NO₃
Molecular Weight: 157,169 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08274999
CAS-Nummer: 60298-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR