6-Bromo-2-methylquinoline-3-carboxylic acid ≥98%

Lieferant: Apollo Scientific
Achtung

OR309203-250MG
APOSOR309203-250MGEA 92.8 EUR
APOSOR309203-250MG
6-Bromo-2-methylquinoline-3-carboxylic acid ≥98%
6-Bromo-2-methylquinoline-3-carboxylic acid
Formel: C₁₁H₈BrNO₂
Molecular Weight: 266,091 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787811
CAS-Nummer: 92513-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR