6-Bromo-2-methylquinoline-3-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

Synonyme: Ethyl 6-bromo-2-methylquinoline-3-carboxylate

OR309204-1G
APOSOR309204-1GEA 287 EUR
APOSOR309204-1G
6-Bromo-2-methylquinoline-3-carboxylic acid ethyl ester
6-Bromo-2-methylquinoline-3-carboxylic acid ethyl ester
Formel: C₁₃H₁₂BrNO₂
Molecular Weight: 294.144 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787812
CAS-Nummer: 948289-14-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR