6-Chloro-4-hydroxy-8-methylquinoline-3-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

Synonyme: Ethyl 6-chloro-4-hydroxy-8-methylquinoline-3-carboxylate

OR309209-1G
APOSOR309209-1GEA 91.5 EUR
APOSOR309209-1G
6-Chloro-4-hydroxy-8-methylquinoline-3-carboxylic acid ethyl ester
6-Chloro-4-hydroxy-8-methylquinoline-3-carboxylic acid ethyl ester
Formel: C₁₃H₁₂ClNO₃
Molecular Weight: 265.692 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173359
CAS-Nummer: 228728-86-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR