6-Chloro-8-methylquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

OR309211-1G
APOSOR309211-1GEA 335 EUR
APOSOR309211-1G
6-Chloro-8-methylquinoline-3-carboxylic acid
6-Chloro-8-methylquinoline-3-carboxylic acid
Formel: C₁₁H₈ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09787823
CAS-Nummer: 948289-56-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR