6-Ethoxy-4-hydrazino-2-methylquinoline hydrochloride

Lieferant: Apollo Scientific

OR309213-1G
APOSOR309213-1GEA 359 EUR
APOSOR309213-1G
6-Ethoxy-4-hydrazino-2-methylquinoline hydrochloride
6-Ethoxy-4-hydrazino-2-methylquinoline hydrochloride
Formel: C₁₂H₁₅N₃O·ClH
Molecular Weight: 253,728 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787829
CAS-Nummer: 1071572-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR