6-Ethyl-4-hydroxyquinoline

Lieferant: Apollo Scientific

Synonyme: 6-Ethylquinolin-4-ol

OR309214-1G
APOSOR309214-1GEA 95.7 EUR
APOSOR309214-1G
6-Ethyl-4-hydroxyquinoline
6-Ethyl-4-hydroxyquinoline
Formel: C₁₁H₁₁NO
Molecular Weight: 173.211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02332229
CAS-Nummer: 303121-13-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR