Ethyl-6-methoxy-2-methylquinoline-3-carboxylate

Lieferant: Apollo Scientific

Synonyme: 6-Methoxy-2-methylquinoline-3-carboxylic acid ethyl ester, Ethyl 6-methoxy-2-methylquinoline-3-carboxylate

OR309216-1G
APOSOR309216-1GEA 346 EUR
APOSOR309216-1G
Ethyl-6-methoxy-2-methylquinoline-3-carboxylate
Ethyl-6-methoxy-2-methylquinoline-3-carboxylate
Formel: C₁₄H₁₅NO₃
Molecular Weight: 245.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01604297
CAS-Nummer: 86210-92-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR