7-Bromo-2-methylquinoline-3-carboxylic acid
Lieferant: Apollo Scientific
OR309221-1G
APOSOR309221-1GEA
383
EUR
APOSOR309221-1G
7-Bromo-2-methylquinoline-3-carboxylic acid
7-Bromo-2-methylquinoline-3-carboxylic acid
Formel:
C₁₁H₈BrNO₂ Molecular Weight: 266,091 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06254448 CAS-Nummer: 610277-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...