7-Bromo-2-methylquinoline-3-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

OR309222-1G
APOSOR309222-1GEA 351 EUR
APOSOR309222-1G
7-Bromo-2-methylquinoline-3-carboxylic acid ethyl ester
7-Bromo-2-methylquinoline-3-carboxylic acid ethyl ester
Formel: C₁₃H₁₂BrNO₂
Molecular Weight: 294,144 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787848
CAS-Nummer: 948290-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR