7-Bromo-4-chloro-2-methylquinoline

Lieferant: Apollo Scientific

OR309223-1G
APOSOR309223-1GEA 348 EUR
APOSOR309223-1G
7-Bromo-4-chloro-2-methylquinoline
7-Bromo-4-chloro-2-methylquinoline
Formel: C₁₀H₇BrClN
Molecular Weight: 256,526 g/mol
Schmelzpunkt: 104…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09787849
CAS-Nummer: 143946-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR