7-Chloro-8-methylquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

OR309225-1G
APOSOR309225-1GEA 335 EUR
APOSOR309225-1G
7-Chloro-8-methylquinoline-3-carboxylic acid
7-Chloro-8-methylquinoline-3-carboxylic acid
Formel: C₁₁H₈ClNO₂
Molecular Weight: 221,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787859
CAS-Nummer: 948290-52-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR