7-Methoxychinaldin

Lieferant: Apollo Scientific

OR309227-1G
APOSOR309227-1GEA 125 EUR
APOSOR309227-1G
7-Methoxychinaldin
7-Methoxychinaldin
Formel: C₁₁H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD09787870
CAS-Nummer: 19490-87-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR