4-Hydroxy-7-methylquinoline

Lieferant: Apollo Scientific

OR309230-1G
APOSOR309230-1GEA 352 EUR
APOSOR309230-1G
4-Hydroxy-7-methylquinoline
4-Hydroxy-7-methylquinoline
Formel: C₁₀H₉NO
Molecular Weight: 159,185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787874
CAS-Nummer: 82121-08-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR