8-Bromo-4-hydroxy-2-methylquinoline

Lieferant: Apollo Scientific

Synonyme: 8-Bromo-2-methylquinolin-4-ol

OR309232-5G
APOSOR309232-5GEA 180 EUR
APOSOR309232-5G
8-Bromo-4-hydroxy-2-methylquinoline
8-Bromo-4-hydroxy-2-methylquinoline
Formel: C₁₀H₈BrNO
Molecular Weight: 238.081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00484434
CAS-Nummer: 1201-08-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR