Ethyl-8-chlor-4-hydroxychinolin-3-carboxylat

Lieferant: Apollo Scientific

Synonyme: Ethyl 8-chloro-4-hydroxyquinoline-3-carboxylate, 8-Chloro-4-hydroxyquinoline-3-carboxylic acid ethyl ester

OR309235-5G
APOSOR309235-5GEA 66.7 EUR
APOSOR309235-5G
Ethyl-8-chlor-4-hydroxychinolin-3-carboxylat
Ethyl-8-chlor-4-hydroxychinolin-3-carboxylat
Formel: C₁₂H₁₀ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00024002
CAS-Nummer: 73987-37-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR