6-Ethoxy-4-hydroxyquinoline

Lieferant: Apollo Scientific

Synonyme: 6-Ethoxyquinolin-4-ol

OR309240-5G
APOSOR309240-5GEA 173 EUR
APOSOR309240-5G
6-Ethoxy-4-hydroxyquinoline
6-Ethoxy-4-hydroxyquinoline
Formel: C₁₁H₁₁NO₂
Molecular Weight: 189.211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02332227
CAS-Nummer: 303121-11-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR