4-Brom-6-chlor-8-methylchinolin

Lieferant: Apollo Scientific

OR309243-1G
APOSOR309243-1GEA 270 EUR
APOSOR309243-1G
4-Brom-6-chlor-8-methylchinolin
4-Bromo-6-chloro-8-methylquinoline
Formel: C₁₀H₇BrClN
Molecular Weight: 256,526 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07700235
CAS-Nummer: 927800-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR