4-Brom-6-methoxy-2-methylchinolin
Lieferant: Apollo Scientific
OR309244-1G
APOSOR309244-1GEA
359
EUR
APOSOR309244-1G
4-Brom-6-methoxy-2-methylchinolin
4-Bromo-6-methoxy-2-methylquinoline
Formel:
C₁₁H₁₀BrNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD07700256 CAS-Nummer: 856095-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...