4-Brom-6-methoxychinolin
Lieferant: Apollo Scientific
OR309245-1G
APOSOR309245-1GEA
46.4
EUR
APOSOR309245-1G
4-Brom-6-methoxychinolin
4-Bromo-6-methoxyquinoline
Formel:
C₁₀H₈BrNO Molecular Weight: 238,0800 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08437074 CAS-Nummer: 42881-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...