4-Hydroxy-2-methylquinoline-6-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

Synonyme: Ethyl 4-hydroxy-2-methylquinoline-6-carboxylate

OR309246-1G
APOSOR309246-1GEA 214 EUR
APOSOR309246-1G
4-Hydroxy-2-methylquinoline-6-carboxylic acid ethyl ester
4-Hydroxy-2-methylquinoline-6-carboxylic acid ethyl ester
Formel: C₁₃H₁₃NO₃
Molecular Weight: 231.247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00440392
CAS-Nummer: 300590-94-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR