4-Piperazin-1-yl-quinoline dihydrochloride

Lieferant: Apollo Scientific

OR309247-1G
APOSOR309247-1GEA 517 EUR
APOSOR309247-1G
4-Piperazin-1-yl-quinoline dihydrochloride
4-Piperazin-1-yl-quinoline dihydrochloride
Formel: C₁₃H₁₅N₃·2ClH
Molecular Weight: 286,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26383403
CAS-Nummer: 1350361-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR