6-Bromo-4-hydroxy-2-methylquinoline

Lieferant: Apollo Scientific

Synonyme: 6-Bromo-2-methylquinolin-4-ol

OR309248-5G
APOSOR309248-5GEA 128 EUR
APOSOR309248-5G
6-Bromo-4-hydroxy-2-methylquinoline
6-Bromo-4-hydroxy-2-methylquinoline
Formel: C₁₀H₈BrNO
Molecular Weight: 238.081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00484433
CAS-Nummer: 103030-28-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR