6-Bromo-4-(piperazin-1-yl)quinoline

Lieferant: Apollo Scientific

OR309249-1G
APOSOR309249-1GEA 383 EUR
APOSOR309249-1G
6-Bromo-4-(piperazin-1-yl)quinoline
6-Bromo-4-(piperazin-1-yl)quinoline
Formel: C₁₃H₁₄BrN₃
Molecular Weight: 292,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03618501
CAS-Nummer: 474707-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR