6-Methoxyquinoline-3-carboxamide

Lieferant: Apollo Scientific

OR309250-1G
APOSOR309250-1GEA 467 EUR
APOSOR309250-1G
6-Methoxyquinoline-3-carboxamide
6-Methoxyquinoline-3-carboxamide
Formel: C₁₁H₁₀N₂O₂
Molecular Weight: 202,209 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07700276
CAS-Nummer: 71083-30-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR