7-Methoxy-4-(piperazin-1-yl)quinoline

Lieferant: Apollo Scientific

OR309252-1G
APOSOR309252-1GEA 250 EUR
APOSOR309252-1G
7-Methoxy-4-(piperazin-1-yl)quinoline
7-Methoxy-4-(piperazin-1-yl)quinoline
Formel: C₁₄H₁₇N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD02179804
CAS-Nummer: 4038-97-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR