6-Chloro-2-hydroxyquinoline-3-carbaldehyde

Lieferant: Apollo Scientific

OR309254-1G
APOSOR309254-1GEA 368 EUR
APOSOR309254-1G
6-Chloro-2-hydroxyquinoline-3-carbaldehyde
6-Chloro-2-hydroxyquinoline-3-carbaldehyde
Formel: C₁₀H₆ClNO₂
Molecular Weight: 207,613 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09881441
CAS-Nummer: 73568-44-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR