7-Chloro-2-hydroxyquinoline-3-carbaldehyde

Lieferant: Apollo Scientific

OR309255-1G
APOSOR309255-1GEA 277 EUR
APOSOR309255-1G
7-Chloro-2-hydroxyquinoline-3-carbaldehyde
7-Chloro-2-hydroxyquinoline-3-carbaldehyde
Formel: C₁₀H₆ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09881442
CAS-Nummer: 73568-43-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR