2,6-Dichloroquinoline-3-methanol

Lieferant: Apollo Scientific

OR309256-1G
APOSOR309256-1GEA 343 EUR
APOSOR309256-1G
2,6-Dichloroquinoline-3-methanol
2,6-Dichloroquinoline-3-methanol
Formel: C₁₀H₇Cl₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD09997988
CAS-Nummer: 1017429-35-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR